Assay Details
Target Gene Details
Entrez Gene ID: | 74053 |
Gene Name: | glutamate receptor interacting protein 1 |
Gene Aliases: |
4931400F03Rik, GRIP, eb |
Location: |
Chr.10:119453835-120087267 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 18 - Exon 19 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Grip1 | NM_001277292.1 | NP_001264221.1 | ||
NM_001277293.1 | NP_001264222.1 | |||
NM_001277294.1 | NP_001264223.1 | |||
NM_001277295.1 | NP_001264224.1 | |||
NM_028736.2 | NP_083012.1 | |||
NM_130891.2 | NP_570961.1 | |||
NM_133442.2 | NP_597699.1 | |||
NR_102387.1 | ||||
AB051560.1 | BAB46929.1 | |||
AB051561.1 | BAB46930.1 | |||
AB051562.1 | BAB46931.1 | |||
AK016420.1 | ||||
AK029845.1 | ||||
AK029905.1 | BAC26668.1 | |||
AK220553.1 | BAD90538.1 | |||
AY255674.1 | AAP81257.1 | |||
AY294283.1 | AAQ56581.1 | |||
BC067398.1 | AAH67398.1 | |||
BC072632.1 | AAH72632.1 |