Assay Details
Target Gene Details
Entrez Gene ID: | 57764 |
Gene Name: | netrin 4 |
Gene Aliases: |
- |
Location: |
Chr.10:93641049-93745972 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ntn4 | NM_021320.3 | 4 | 1324 | NP_067295.2 |
AF268066.1 | 4 | 919 | AAF91404.1 | |
AF281278.1 | 4 | 1172 | AAG30823.1 | |
BC119182.1 | 4 | 1006 | ||
BC119184.1 | 4 | 1006 | ||
CX205916.1 | 2 | 135 |
More Information
Set Membership: |
Intragenic Exonic |