Assay Details
Target Gene Details
Entrez Gene ID: | 74782 |
Gene Name: | glycosyltransferase 8 domain containing 2 |
Gene Aliases: |
1110021D20Rik |
Location: |
Chr.10:82650428-82694113 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Glt8d2 | NM_029102.3 | NP_083378.2 | ||
AK003894.1 | BAB23061.1 | |||
AK040842.1 | BAC30718.1 | |||
AK086276.1 | 2 | 221 | ||
AK136351.1 | ||||
BC082561.1 | AAH82561.1 |
More Information
Set Membership: |
Intragenic Non-exonic |