Assay Details
Target Gene Details
Entrez Gene ID: | 103199 |
Gene Name: | FIG4 homolog (S. cerevisiae) |
Gene Aliases: |
A530089I17Rik, AI326867, Sac3 |
Location: |
Chr.10:41188172-41303241 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Fig4 | NM_133999.1 | 14 | 1702 | NP_598760.1 |
AK041196.1 | 14 | 1703 | ||
AK172130.1 | 14 | 1696 | BAE42839.1 | |
AK172341.1 | 14 | 1667 | ||
AK172926.1 | 7 | 750 | BAD32204.1 | |
AK181110.1 | 3 | 166 | ||
AK204518.1 | 3 | 230 | ||
BC015295.1 | 14 | 1702 | AAH15295.1 | |
BC031887.1 | 14 | 1701 | AAH31887.1 |
More Information
Set Membership: |
Intragenic Exonic |