Assay Details
Target Gene Details
Entrez Gene ID: | 209558 |
Gene Name: | ectonucleotide pyrophosphatase/phosphodiesterase 3 |
Gene Aliases: |
AI876438, CD203c |
Location: |
Chr.10:24773814-24836195 on Build GRCm38 |
Assay Gene Location: | Within Exon 18 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Enpp3 | NM_134005.2 | 18 | 1713 | NP_598766.2 |
AK132256.1 | 18 | 1726 | ||
AY630402.1 | 18 | 1611 | AAT64421.1 | |
BC099535.1 | 4 | 315 | AAH99535.1 |