Assay Details
Target Gene Details
Entrez Gene ID: | 71567 |
Gene Name: | minichromosome maintenance complex component 9 |
Gene Aliases: |
9030408O17Rik, Gm235, Mcmdc1 |
Location: |
Chr.10:53536315-53630439 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mcm9 | NM_027830.2 | NP_082106.2 | ||
AK018494.1 | BAB31238.1 | |||
AK046636.1 | BAE20656.1 | |||
AK139112.1 |
More Information
Set Membership: |
Intragenic Non-exonic |