Assay Details
Target Gene Details
Entrez Gene ID: | 94221 |
Gene Name: | golgi associated PDZ and coiled-coil motif containing |
Gene Aliases: |
2210402P09Rik, AI844555, CAL, FIG, GOPC1, PIST |
Location: |
Chr.10:52337024-52382124 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gopc | NM_001199272.1 | NP_001186201.1 | ||
NM_053187.3 | NP_444417.2 | |||
AB052838.1 | BAB69946.1 | |||
AF287893.1 | AAG00571.1 | |||
AK030427.1 | BAC26958.1 | |||
AK030637.1 | BAC27058.1 | |||
AK032613.1 | BAC27951.1 | |||
AK083232.1 | 1 | 1175 | ||
BC051171.1 | AAH51171.1 |
More Information
Set Membership: |
Intragenic Non-exonic |