Assay Details
Target Gene Details
Entrez Gene ID: | 13139 |
Gene Name: | diacylglycerol kinase, alpha |
Gene Aliases: |
80kDa, AW146112, Dagk1 |
Location: |
Chr.10:128720134-128748420 on Build GRCm38 |
Assay Gene Location: | Within Intron 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Dgka | NM_016811.2 | NP_058091.2 | ||
AF085219.1 | AAC33483.1 | |||
AK008063.1 | ||||
AK008272.1 | ||||
AK008314.1 | ||||
AK037536.1 | 1 | 916 | ||
BC006713.1 | AAH06713.1 | |||
JN832677.1 | ||||
JN832678.1 | ||||
JN832679.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic |