Assay Details
Target Gene Details
Entrez Gene ID: | 108853 |
Gene Name: | mitochondrial translational release factor 1-like |
Gene Aliases: |
9130004K12Rik |
Location: |
Chr.10:5811119-5824390 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mtrf1l | NM_175374.3 | NP_780583.1 | ||
AK036424.1 | BAE20502.1 | |||
AK078897.1 | BAC37446.1 | |||
AK185802.1 |
More Information
Set Membership: |
Intragenic Non-exonic |