Assay Details
Target Gene Details
Entrez Gene ID: | 171580 |
Gene Name: | microtubule associated monooxygenase, calponin and LIM domain containing 1 |
Gene Aliases: |
MICAL, MICAL-1, Nical |
Location: |
Chr.10:41475860-41487030 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 21 - Exon 22 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mical1 | NM_001164433.1 | NP_001157905.1 | ||
NM_138315.2 | NP_612188.1 | |||
AK154665.1 | ||||
AK154930.1 | ||||
AK157196.1 | ||||
AK159369.1 | BAE35027.1 | |||
AK170382.1 | ||||
AK170827.1 | ||||
AK170874.1 | ||||
AK170881.1 | BAE42090.1 | |||
AK171234.1 | BAE42330.1 | |||
AK171429.1 | BAE42447.1 | |||
AK171479.1 | BAE42481.1 | |||
AK172562.1 | ||||
AK172716.1 | ||||
AK182215.1 | ||||
AK186327.1 | ||||
BC021477.1 | AAH21477.1 | |||
BC034682.1 | AAH34682.1 |