Assay Details
Target Gene Details
Entrez Gene ID: | 216445 |
Gene Name: | Rho GTPase activating protein 9 |
Gene Aliases: |
AU043488 |
Location: |
Chr.10:127321959-127329946 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Arhgap9 | NM_001285785.1 | NP_001272714.1 | ||
NM_146011.2 | NP_666123.2 | |||
AK047280.1 | BAC33014.1 | |||
AK049245.1 | BAC33633.1 | |||
AK080955.1 | 6 | 1586 | BAC38093.1 | |
AK083474.1 | ||||
AK089051.1 | BAC40720.1 | |||
AK167738.1 | BAE39777.1 | |||
BC024535.1 | ||||
BC027374.1 | ||||
DQ498128.1 |
More Information
Set Membership: |
Intragenic Non-exonic |