Assay Details
Target Gene Details
Entrez Gene ID: | 13198 |
Gene Name: | DNA-damage inducible transcript 3 |
Gene Aliases: |
CHOP-10, CHOP10, chop, gadd153 |
Location: |
Chr.10:127290793-127296288 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ddit3 | NM_001290183.1 | NP_001277112.1 | ||
NM_007837.4 | NP_031863.3 | |||
AA929408.1 | ||||
AK028120.1 | BAE20435.1 | |||
BC013718.1 | AAH13718.1 | |||
BE134148.1 | ||||
BY121546.1 | ||||
X67083.1 | CAA47465.1 |