Assay Details
Target Gene Details
Entrez Gene ID: | 216001 |
Gene Name: | mitochondrial calcium uptake 1 |
Gene Aliases: |
C730016L05Rik, Calc, Cbara1 |
Location: |
Chr.10:59702482-59864134 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Micu1 | NM_001291442.1 | 4 | 581 | NP_001278371.1 |
NM_001291443.1 | 4 | 581 | NP_001278372.1 | |
NM_144822.3 | 4 | 581 | NP_659071.1 | |
AK046049.1 | 4 | 546 | BAC32581.1 | |
AK049521.1 | 4 | 577 | BAC33792.1 | |
AK050113.1 | 4 | 568 | BAC34073.1 | |
AK051470.1 | 4 | 487 | ||
AK052485.1 | 4 | 529 | BAC35013.1 | |
AK077605.1 | 4 | 531 | BAC36895.1 | |
AK083520.1 | 4 | 584 | ||
AK085553.1 | 4 | 497 | ||
AK189367.1 | ||||
AK200391.1 | ||||
BC018320.1 | 4 | 522 | AAH18320.1 | |
BC023022.1 | 4 | 516 | AAH23022.1 | |
BC023042.1 | 3 | 465 | AAH23042.1 | |
BC026566.1 | 4 | 532 | AAH26566.1 |
More Information
Set Membership: |
Intragenic Exonic |