Assay Details
Target Gene Details
Entrez Gene ID: | 66686 |
Gene Name: | discoidin, CUB and LCCL domain containing 1 |
Gene Aliases: |
4631413K11Rik |
Location: |
Chr.10:52233584-52321378 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Dcbld1 | NM_025705.3 | 2 | 319 | NP_079981.2 |
AK014521.1 | 2 | 131 | BAB29409.1 | |
AK016485.1 | 2 | 319 | BAB30265.1 | |
AK157211.1 | 2 | 306 | ||
BC026771.1 | AAH26771.1 | |||
BY096153.1 |
More Information
Set Membership: |
Intragenic Exonic |