Assay Details
Target Gene Details
Entrez Gene ID: | 11783 |
Gene Name: | apoptotic peptidase activating factor 1 |
Gene Aliases: |
6230400I06Rik, Apaf-1, Apaf1l, fog, mKIAA0413 |
Location: |
Chr.10:90989311-91082743 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Apaf1 | NM_001042558.1 | 6 | 1435 | NP_001036023.1 |
NM_001282947.1 | 6 | 1359 | NP_001269876.1 | |
NM_009684.2 | 6 | 1392 | NP_033814.2 | |
AF064071.1 | 6 | 1319 | AAC62458.1 | |
AK220340.1 | 6 | 1431 | BAD90406.1 | |
BC131683.1 | 6 | 951 | ||
BC131684.1 | 6 | 951 |