Assay Details
Target Gene Details
Entrez Gene ID: | 12461 |
Gene Name: | chaperonin containing Tcp1, subunit 2 (beta) |
Gene Aliases: |
Cctb |
Location: |
Chr.10:117050997-117063814 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cct2 | NM_007636.2 | 7 | 698 | NP_031662.2 |
AB041570.1 | 7 | 670 | BAA95054.1 | |
AK012295.1 | 7 | 654 | ||
AK075581.1 | 7 | 671 | BAC35834.1 | |
AK088145.1 | 7 | 660 | ||
AK145406.1 | 7 | 699 | BAE26419.1 | |
AK166891.1 | 7 | 671 | BAE39096.1 | |
AK179930.1 | ||||
BC007470.1 | 7 | 662 | AAH07470.1 | |
BC026918.1 | 7 | 664 | AAH26918.1 | |
Z31553.1 | 7 | 649 | CAA83428.1 |
More Information
Set Membership: |
Intragenic Exonic |