Assay Details
Target Gene Details
Entrez Gene ID: | 12461 |
Gene Name: | chaperonin containing Tcp1, subunit 2 (beta) |
Gene Aliases: |
Cctb |
Location: |
Chr.10:117050997-117063814 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cct2 | NM_007636.2 | NP_031662.2 | ||
AB041570.1 | BAA95054.1 | |||
AK012295.1 | ||||
AK075581.1 | BAC35834.1 | |||
AK088145.1 | ||||
AK145406.1 | BAE26419.1 | |||
AK166891.1 | BAE39096.1 | |||
AK179930.1 | ||||
BC007470.1 | AAH07470.1 | |||
BC026918.1 | AAH26918.1 | |||
Z31553.1 | CAA83428.1 |
More Information
Set Membership: |
Intragenic Non-exonic |