Assay Details
Target Gene Details
Entrez Gene ID: | 27403 |
Gene Name: | ATP-binding cassette, sub-family A (ABC1), member 7 |
Gene Aliases: |
ABCX, Abc51 |
Location: |
Chr.10:79996453-80015572 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Abca7 | NM_013850.1 | NP_038878.1 | ||
AF287141.1 | AAK56862.1 | |||
BC024511.1 | AAH24511.1 | |||
BC145120.1 | ||||
BC150708.1 |
More Information
Set Membership: |
Intragenic Non-exonic |