Assay Details
Target Gene Details
Entrez Gene ID: | 215821 |
Gene Name: | ARFGEF family member 3 |
Gene Aliases: |
9630004D11, B930094H20Rik, BIG3, D10Bwg1379e, mKIAA1244 |
Location: |
Chr.10:18581839-18744069 on Build GRCm38 |
Assay Gene Location: | Within Exon 29 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Arfgef3 | NM_001033258.4 | 29 | 4829 | NP_001028430.1 |
AK035782.1 | 6 | 741 | ||
AK122475.1 | 18 | 3191 | BAC65757.1 | |
AK147679.1 | 29 | 4830 | BAE28069.1 | |
AK147963.1 | 29 | 4827 | ||
AK148343.1 | 29 | 4774 | ||
AK165602.1 | 29 | 4771 |
More Information
Set Membership: |
Intragenic Exonic |