Assay Details
Target Gene Details
Entrez Gene ID: | 77531 |
Gene Name: | ankyrin repeat and sterile alpha motif domain containing 1B |
Gene Aliases: |
AIDA-1b, C030032C09Rik, E530015N03Rik, ENSMUSG00000078431, Gm10937, Gm1555 |
Location: |
Chr.10:89873509-90973300 on Build GRCm38 |
Assay Gene Location: | Within Exon 17 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Anks1b | NM_001128086.2 | 17 | 2842 | NP_001121558.2 |
NM_001177396.1 | 3 | 673 | NP_001170867.1 | |
NM_001177397.1 | 3 | 288 | NP_001170868.1 | |
NM_001177398.1 | 3 | 673 | NP_001170869.1 | |
NM_181398.3 | 3 | 673 | NP_852063.1 | |
AK032211.1 | 3 | 675 | BAC27761.1 | |
AK034154.1 | 4 | 308 | BAC28610.1 | |
AK049899.1 | 3 | 618 | BAC33978.1 | |
AK132228.1 | 3 | 640 | ||
AK163750.1 | 3 | 647 | BAE37478.1 | |
BC098373.1 | 3 | 288 | AAH98373.1 |
More Information
Set Membership: |
Intragenic Exonic |