Assay Details
Target Gene Details
Entrez Gene ID: | 235028 |
Gene Name: | zinc finger protein 426 |
Gene Aliases: |
2900057C04Rik, KRAB1, Zfo61, Zfp68-rs1, Znf426, mKIAA4237 |
Location: |
Chr.9:20468548-20493922 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Zfp426 | NM_001110309.1 | NP_001103779.1 | ||
NM_146221.4 | NP_666333.1 | |||
AK036912.1 | BAC29636.1 | |||
AK085144.1 | BAC39374.1 | |||
AK220563.1 | BAD90329.1 | |||
BC023931.1 | AAH23931.1 | |||
BC024805.1 | AAH24805.1 | |||
U29495.1 | AAC52290.1 |
More Information
Set Membership: |
Intragenic Non-exonic |