Assay Details
Target Gene Details
Entrez Gene ID: | 235628 |
Gene Name: | protease, serine 42 |
Gene Aliases: |
Tessp2 |
Location: |
Chr.9:110797261-110803744 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Prss42 | NM_153099.1 | 4 | 760 | NP_694739.1 |
AB052292.1 | 4 | 760 | BAB78735.1 | |
AK133402.1 | 4 | 625 | BAE21636.1 | |
BC060984.1 | 4 | 779 | AAH60984.1 |
More Information
Set Membership: |
Intragenic Exonic |