Assay Details
Target Gene Details
Entrez Gene ID: | 100038566 |
Gene Name: | predicted gene 10658 |
Gene Aliases: |
ENSMUSG00000074284 |
Location: |
Chr.9:57056897-57071966 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gm10658 | NR_045886.1 | |||
AK085949.1 | ||||
AK143982.1 | BAE25644.1 | |||
AK162906.1 | BAE37109.1 | |||
BU696022.1 |
Target Gene Details
Entrez Gene ID: | 56294 |
Gene Name: | protein tyrosine phosphatase, non-receptor type 9 |
Gene Aliases: |
MEG2 |
Location: |
Chr.9:56994968-57062808 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ptpn9 | NM_019651.2 | NP_062625.2 | ||
AF013490.1 | AAB66898.1 | |||
AK089069.1 | ||||
AK191509.1 | ||||
BC053017.1 | AAH53017.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic |