Assay Details
Target Gene Details
Entrez Gene ID: | 14012 |
Gene Name: | myelin protein zero-like 2 |
Gene Aliases: |
Eva, Eva1 |
Location: |
Chr.9:45040291-45054043 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mpzl2 | NM_007962.4 | 4 | 940 | NP_031988.2 |
AF030454.1 | 4 | 721 | AAC40128.1 | |
AK032788.1 | 4 | 721 | BAC28022.1 | |
AK166341.1 | 4 | 671 | BAE38716.1 | |
AK167263.1 | 4 | 704 | BAE39377.1 | |
AK170816.1 | 4 | 706 | BAE42048.1 | |
BC015076.1 | 4 | 766 | AAH15076.1 |
More Information
Set Membership: |
Intragenic Exonic |