Assay Details
Target Gene Details
Entrez Gene ID: | 64011 |
Gene Name: | neurogranin |
Gene Aliases: |
0710001B06Rik, AI838505, NG, NG/RC3, Pss1, R75334, RC3 |
Location: |
Chr.9:37544493-37552745 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Nrgn | NM_022029.2 | NP_071312.1 | ||
AK002933.1 | BAB22466.1 | |||
AK158630.1 | BAE34589.1 | |||
BC061102.1 | AAH61102.1 | |||
BC138511.1 | ||||
BC138513.1 | ||||
BU604757.1 |