Assay Details
Target Gene Details
Entrez Gene ID: | 319239 |
Gene Name: | neuropeptide S receptor 1 |
Gene Aliases: |
9330128H10Rik, GPRA, Gpr154, MVTR, PGR14, VRR1 |
Location: |
Chr.9:24097950-24317590 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Npsr1 | NM_175678.3 | NP_783609.1 | ||
AK033957.1 | BAC28526.1 | |||
AK041451.1 | BAC30949.1 | |||
AY299387.1 |
More Information
Set Membership: |
Intragenic Non-exonic |