Assay Details
Target Gene Details
Entrez Gene ID: | 12145 |
Gene Name: | chemokine (C-X-C motif) receptor 5 |
Gene Aliases: |
Blr1, CXC-R5, CXCR-5, Gpcr6, MDR15 |
Location: |
Chr.9:44511534-44561874 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cxcr5 | NM_007551.2 | NP_031577.2 | ||
AK089611.1 | ||||
AK133064.1 | BAE21492.1 | |||
AK155954.1 | BAE33519.1 | |||
AK156087.1 | BAE33583.1 | |||
AK157016.1 | BAE33932.1 | |||
BC064059.1 | AAH64059.1 |