Assay Details
Target Gene Details
Entrez Gene ID: | 13036 |
Gene Name: | cathepsin H |
Gene Aliases: |
AL022844 |
Location: |
Chr.9:90054267-90076095 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ctsh | NM_007801.2 | NP_031827.2 | ||
AK017638.1 | ||||
AK149949.1 | BAE29188.1 | |||
AK150583.1 | BAE29677.1 | |||
AK151877.1 | ||||
AK157376.1 | BAE34071.1 | |||
AK160026.1 | BAE35569.1 | |||
AK167498.1 | ||||
AK171550.1 | ||||
BC006878.1 | AAH06878.1 | |||
U06119.1 | AAA82966.1 |
More Information
Set Membership: |
Intragenic Non-exonic |