Assay Details
Target Gene Details
Entrez Gene ID: | 67213 |
Gene Name: | CKLF-like MARVEL transmembrane domain containing 6 |
Gene Aliases: |
2810051A14Rik, AA536733, Cklfsf6 |
Location: |
Chr.9:114731157-114749347 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cmtm6 | NM_026036.3 | 1 | 84 | NP_080312.1 |
AK012941.1 | 1 | 85 | BAB28562.1 | |
AK030885.1 | 1 | 64 | BAC27172.1 | |
AK128960.1 | 1 | 70 | ||
AK145743.1 | 1 | 65 | BAE26621.1 | |
AK150775.1 | 1 | 85 | ||
AK151092.1 | 1 | 85 | ||
AK151431.1 | 1 | 85 | ||
AK152448.1 | 1 | 86 | BAE31227.1 | |
AK152723.1 | 1 | 95 | BAE31446.1 | |
AK152817.1 | 1 | 95 | BAE31520.1 | |
AK153092.1 | 1 | 85 | ||
AK154481.1 | 1 | 71 | BAE32617.1 | |
AK159439.1 | 1 | 54 | BAE35085.1 | |
AK159735.1 | 1 | 79 | BAE35329.1 | |
AK165948.1 | 1 | 53 | BAE38480.1 | |
AK167499.1 | 1 | 71 | BAE39575.1 | |
AK167854.1 | 1 | 76 | BAE39872.1 | |
AK170905.1 | 1 | 86 | BAE42106.1 | |
AK171394.1 | 1 | 115 | BAE42431.1 | |
AY241868.1 | 1 | 121 | AAP33490.1 | |
BC027248.1 | 1 | 56 | AAH27248.1 |
More Information
Set Membership: |
Intragenic Exonic |