Assay Details
Target Gene Details
Entrez Gene ID: | 12776 |
Gene Name: | chemokine (C-C motif) receptor 8 |
Gene Aliases: |
C-C, C-C CKR-8, CC-CKR-8, CCR-8, CKR-8, Cmkbr8, mCCR8 |
Location: |
Chr.9:120092133-120094906 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ccr8 | NM_007720.2 | NP_031746.1 | ||
Z98205.1 | CAB10895.1 |