Assay Details
Target Gene Details
Entrez Gene ID: | 13070 |
Gene Name: | cytochrome P450, family 11, subfamily a, polypeptide 1 |
Gene Aliases: |
Cyp11a, Cypxia1, D9Ertd411e, P450scc, Scc, cscc |
Location: |
Chr.9:58015017-58027023 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cyp11a1 | NM_019779.3 | NP_062753.3 | ||
AF195119.1 | AAF03897.1 | |||
AK077945.1 | ||||
BC068264.1 | AAH68264.1 |