Assay Details
Target Gene Details
Entrez Gene ID: | 67023 |
Gene Name: | unconventional SNARE in the ER 1 homolog (S. cerevisiae) |
Gene Aliases: |
2010315L10Rik, 5730403H22Rik, AV002165, D12, Ed2, Q-snare, p31 |
Location: |
Chr.8:71366848-71369732 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Use1 | NM_001145780.1 | 9 | 964 | NP_001139252.1 |
NM_025917.4 | 8 | 751 | NP_080193.1 | |
AF353245.1 | 8 | 744 | AAK26661.1 | |
AK003257.1 | 8 | 744 | ||
AK003385.1 | 7 | 615 | ||
AK008573.1 | 8 | 744 | BAB25752.1 | |
AK008661.1 | 7 | 649 | ||
AK012490.1 | 8 | 745 | BAB28275.1 | |
AK017484.1 | 5 | 818 | BAB30767.1 | |
AK160726.1 | 8 | 744 | BAE35971.1 | |
AK165639.1 | 8 | 893 | ||
AY842453.1 | 8 | 730 | AAW32097.1 | |
BC025086.1 | 7 | 827 | ||
BC038519.1 | 7 | 814 | AAH38519.1 | |
BC075695.1 | 9 | 870 | AAH75695.1 |