Assay Details
Target Gene Details
Entrez Gene ID: | 67023 |
Gene Name: | unconventional SNARE in the ER 1 homolog (S. cerevisiae) |
Gene Aliases: |
2010315L10Rik, 5730403H22Rik, AV002165, D12, Ed2, Q-snare, p31 |
Location: |
Chr.8:71366848-71369732 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Use1 | NM_001145780.1 | 2 | 290 | NP_001139252.1 |
NM_025917.4 | 1 | 77 | NP_080193.1 | |
AF353245.1 | 1 | 70 | AAK26661.1 | |
AK003257.1 | 1 | 70 | ||
AK008573.1 | 1 | 70 | BAB25752.1 | |
AK012490.1 | 1 | 71 | BAB28275.1 | |
AK160726.1 | 1 | 70 | BAE35971.1 | |
AK165639.1 | 1 | 73 | ||
AY842453.1 | 1 | 56 | AAW32097.1 | |
BC025086.1 | 1 | 77 | ||
BC038519.1 | 1 | 61 | AAH38519.1 | |
BC075695.1 | 2 | 196 | AAH75695.1 | |
BY097923.1 | 2 | 291 | ||
BY286283.1 |