Assay Details
Target Gene Details
Entrez Gene ID: | 67023 |
Gene Name: | unconventional SNARE in the ER 1 homolog (S. cerevisiae) |
Gene Aliases: |
2010315L10Rik, 5730403H22Rik, AV002165, D12, Ed2, Q-snare, p31 |
Location: |
Chr.8:71366848-71369732 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Use1 | NM_001145780.1 | 1 | 156 | NP_001139252.1 |
BC075695.1 | 1 | 62 | AAH75695.1 | |
BY097923.1 | 1 | 157 | ||
BY286283.1 | 1 | 61 |