Assay Details
Target Gene Details
Entrez Gene ID: | 11739 |
Gene Name: | solute carrier family 25 (mitochondrial carrier, adenine nucleotide translocator), member 4 |
Gene Aliases: |
AU019225, Ant1 |
Location: |
Chr.8:46207339-46211009 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Slc25a4 | NM_007450.4 | NP_031476.3 | ||
AK051172.1 | ||||
AK078077.1 | BAC37117.1 | |||
AK177376.1 | ||||
BC003791.1 | AAH03791.1 | |||
BC026925.1 | AAH26925.1 | |||
U27315.1 | AAC52837.1 | |||
X74510.1 | CAA52616.1 |
More Information
Set Membership: |
Intragenic Non-exonic |