Assay Details
Target Gene Details
Entrez Gene ID: | 234724 |
Gene Name: | tyrosine aminotransferase |
Gene Aliases: |
- |
Location: |
Chr.8:109990436-109999804 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tat | NM_146214.3 | 6 | 734 | NP_666326.1 |
AK090244.1 | 6 | 732 | BAC41146.1 | |
AK149383.1 | 6 | 732 | BAE28844.1 | |
AK149526.1 | 6 | 732 | BAE28939.1 | |
BC023949.1 | 6 | 718 | AAH23949.1 | |
BC024120.1 | 6 | 709 | AAH24120.1 | |
BC024264.1 | 5 | 657 | AAH24264.1 | |
BC025934.1 | 6 | 669 | AAH25934.1 | |
BC028821.1 | 6 | 709 | AAH28821.1 | |
BC030728.1 | 6 | 713 | AAH30728.1 | |
BC030729.1 | 6 | 707 | AAH30729.1 | |
BC037526.1 | 6 | 689 | AAH37526.1 |
More Information
Set Membership: |
Intragenic Exonic |