Assay Details
Target Gene Details
Entrez Gene ID: | 68219 |
Gene Name: | nudix (nucleoside diphosphate linked moiety X)-type motif 21 |
Gene Aliases: |
25kDa, 3110048P04Rik, 5730530J16Rik, AU014860, AW549947, Cpsf5 |
Location: |
Chr.8:94019403-94037039 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Nudt21 | NM_026623.3 | NP_080899.1 | ||
AK011688.2 | BAB27778.1 | |||
AK012308.1 | BAB28154.1 | |||
AK019433.1 | BAB31718.1 | |||
AK020150.1 | ||||
AK146419.1 | BAE27154.1 | |||
AK160147.1 | BAE35655.1 | |||
AK188064.1 | ||||
AK208594.1 | ||||
AK213941.1 | ||||
BC008270.1 | AAH08270.1 |
More Information
Set Membership: |
Intragenic Non-exonic |