Assay Details
Target Gene Details
Entrez Gene ID: | 66432 |
Gene Name: | solute carrier family 7, member 6 opposite strand |
Gene Aliases: |
2010007L18Rik, 2400002F02Rik |
Location: |
Chr.8:106200438-106210992 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Slc7a6os | NM_001007567.2 | 1 | 119 | NP_001007568.1 |
AK010254.1 | 1 | 110 | ||
AK134626.1 | ||||
AK146733.1 | 1 | 120 | BAE27395.1 | |
AK171156.1 | 1 | 120 | ||
AK171401.1 | 1 | 112 | ||
AK171806.1 | 1 | 122 | ||
BC054118.1 | 1 | 117 | AAH54118.1 |