Assay Details
Target Gene Details
Entrez Gene ID: | 192654 |
Gene Name: | phospholipase A2, group XV |
Gene Aliases: |
ACS, C87498, LLPL, Lpla2, Lypla3 |
Location: |
Chr.8:106150351-106164715 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pla2g15 | NM_133792.2 | NP_598553.1 | ||
AF468958.1 | AAL78651.1 | |||
AK028614.1 | ||||
AK085194.1 | BAC39387.1 | |||
AK155004.1 | BAE32987.1 | |||
AK163111.1 | BAE37197.1 | |||
AK170814.1 | BAE42046.1 | |||
AK180645.1 | ||||
AK186155.1 | ||||
AK188333.1 | ||||
AK189283.1 | ||||
AK203519.1 | ||||
AK205252.1 | ||||
AK205480.1 | ||||
AK207442.1 | ||||
AK211077.1 | ||||
AK212406.1 | ||||
AK212507.1 | ||||
AK212587.1 | ||||
AK212623.1 | ||||
AK214459.1 | ||||
AK215050.1 | ||||
AK215274.1 | ||||
AK215692.1 | ||||
AK215942.1 | ||||
AK217196.1 | ||||
AK219754.1 | ||||
AK220071.1 | ||||
BC019373.1 | AAH19373.1 |
More Information
Set Membership: |
Intragenic Non-exonic |