Assay Details
Target Gene Details
Entrez Gene ID: | 13685 |
Gene Name: | eukaryotic translation initiation factor 4E binding protein 1 |
Gene Aliases: |
4e-bp1, AA959816, PHAS-I |
Location: |
Chr.8:27260327-27275656 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Eif4ebp1 | NM_007918.3 | NP_031944.3 | ||
AK167836.1 | BAE39857.1 | |||
AK169979.1 | BAE41494.1 | |||
AK170031.1 | BAE41521.1 | |||
AK192788.1 | ||||
BC002045.1 | AAH02045.1 | |||
BE651550.1 | ||||
U28656.1 | AAA88818.1 |
More Information
Set Membership: |
Intragenic Non-exonic |