Assay Details
Target Gene Details
Entrez Gene ID: | 17751 |
Gene Name: | metallothionein 3 |
Gene Aliases: |
Mt-3 |
Location: |
Chr.8:94152607-94154148 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mt3 | NM_013603.2 | 3 | 451 | NP_038631.1 |
AK044045.1 | ||||
AK140209.1 | 3 | 452 | BAE24282.1 | |
BC059725.1 | 3 | 349 | AAH59725.1 |