Assay Details
Target Gene Details
Entrez Gene ID: | 234155 |
Gene Name: | membrane bound O-acyltransferase domain containing 4 |
Gene Aliases: |
GOAT, Gm171 |
Location: |
Chr.8:34108219-34125185 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mboat4 | NM_001126314.2 | NP_001119786.1 | ||
BC147844.1 | ||||
BC147849.1 | ||||
EU518496.1 | ||||
EU721729.1 |
More Information
Set Membership: |
Intragenic Non-exonic |