Assay Details
Target Gene Details
Entrez Gene ID: | 16795 |
Gene Name: | like-glycosyltransferase |
Gene Aliases: |
BPFD#36, Gyltl1a, Mbp-1, Mbp1, enr, fg, froggy, mKIAA0609, myd |
Location: |
Chr.8:72814599-73353577 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Large | NM_010687.1 | 11 | 1546 | NP_034817.1 |
AJ006278.1 | 11 | 1546 | CAA06945.1 | |
AK122328.1 | 12 | 2623 | BAC65610.1 | |
AK133940.1 | 11 | 2031 | ||
BC061506.1 | 10 | 1303 | AAH61506.1 | |
BC100399.1 | 11 | 1609 | AAI00400.1 | |
BC125487.1 | 11 | 1512 | ||
BC125489.1 | 11 | 1512 | ||
HM141820.1 | 9 | 1181 |
More Information
Set Membership: |
Intragenic Exonic |