Assay Details
Target Gene Details
Entrez Gene ID: | 52120 |
Gene Name: | heparan-alpha-glucosaminide N-acetyltransferase |
Gene Aliases: |
9430010M12Rik, AW208455, D8Ertd354e, Tmem76 |
Location: |
Chr.8:25944459-25976744 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Hgsnat | NM_029884.1 | 3 | 413 | NP_084160.1 |
AK035264.1 | 3 | 346 | BAC29006.1 | |
AK149883.1 | 3 | 417 | BAE29143.1 | |
AK152926.1 | 3 | 378 | BAE31601.1 | |
AK159649.1 | 3 | 342 | BAE35261.1 | |
AK160068.1 | 3 | 336 | BAE35603.1 | |
AK198139.1 | 1 | 54 | ||
BC024084.1 | 3 | 320 | AAH24084.1 |