Assay Details
Target Gene Details
Entrez Gene ID: | 76615 |
Gene Name: | glutamic-oxaloacetic transaminase 1-like 1 |
Gene Aliases: |
1700083M11Rik |
Location: |
Chr.8:27197459-27223848 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Got1l1 | NM_029674.1 | 1 | 137 | NP_083950.1 |
AK006984.1 | 1 | 124 | BAB24820.1 | |
BC052754.1 | 1 | 137 | AAH52754.1 |
More Information
Set Membership: |
Intragenic Exonic |