Assay Details
Target Gene Details
Entrez Gene ID: | 69165 |
Gene Name: | CD209b antigen |
Gene Aliases: |
1810030I22Rik, DC-SIGNR1, OtB7, SIGNR1, mSIGNR1 |
Location: |
Chr.8:3917165-3926841 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cd209b | NM_001037800.3 | NP_001032889.3 | ||
NM_001287211.1 | NP_001274140.1 | |||
NM_026972.5 | NP_081248.4 | |||
NR_104430.2 | ||||
AF373409.1 | AAL13235.1 | |||
AF374471.1 | AAL27540.1 | |||
AF422108.1 | AAN31450.1 | |||
AF424790.1 | AAN75585.1 | |||
AF424791.1 | AAN75586.1 | |||
AF424792.1 | AAN75587.1 | |||
AF424793.1 | AAN75588.1 | |||
AF424794.1 | AAN75589.1 | |||
AF424795.1 | AAN75590.1 | |||
AF424796.1 | AAN75591.1 | |||
AF424797.1 | AAN75592.1 | |||
AF424798.1 | AAN75593.1 | |||
AF424799.1 | AAN75594.1 | |||
AF424803.1 | AAN75598.1 | |||
AK007656.1 | BAB25166.1 | |||
AK138999.1 | BAE23855.1 |
More Information
Set Membership: |
Intragenic Non-exonic |