Assay Details
Target Gene Details
Entrez Gene ID: | 170786 |
Gene Name: | CD209a antigen |
Gene Aliases: |
CD209, CDSIGN, CIRE, DC-SIGN, DC-SIGN1, Dcsign, SIGN-R1, SIGNR5 |
Location: |
Chr.8:3743395-3748984 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cd209a | NM_133238.5 | NP_573501.1 | ||
AF373408.1 | AAL13234.1 | |||
AF374470.1 | AAL27539.1 | |||
AK054139.1 | BAC35667.1 | |||
AK171639.1 | BAE42579.1 | |||
AY049062.1 | AAK85825.1 | |||
BC111095.1 | AAI11096.1 |
More Information
Set Membership: |
Intragenic Non-exonic |