Assay Details
Target Gene Details
Entrez Gene ID: | 407789 |
Gene Name: | cDNA sequence BC048644 |
Gene Aliases: |
- |
Location: |
Chr.8:121907833-121918428 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
BC048644 | NM_001033485.2 | 1 | 90 | NP_001028657.1 |
AK082595.1 | 1 | 92 | ||
AK133128.1 | 1 | 91 | BAE21521.1 |