Assay Details
Target Gene Details
Entrez Gene ID: | 70993 |
Gene Name: | protease, serine 54 |
Gene Aliases: |
4931432M23Rik, Klkbl4 |
Location: |
Chr.8:95559067-95576330 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Prss54 | NM_027640.1 | NP_081916.1 | ||
AK016501.1 | ||||
BC144859.1 | ||||
BC145787.1 |
More Information
Set Membership: |
Intragenic Non-exonic |