Assay Details
Target Gene Details
Entrez Gene ID: | 13680 |
Gene Name: | DEAD (Asp-Glu-Ala-Asp) box polypeptide 19a |
Gene Aliases: |
DBP5, Ddx19, Eif4a-rs1 |
Location: |
Chr.8:110974556-110997823 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ddx19a | NM_007916.2 | NP_031942.2 | ||
AK049467.1 | BAC33762.1 | |||
AK146548.1 | BAE27252.1 | |||
BC011270.1 | AAH11270.1 | |||
L25125.1 | AAA53629.1 |